missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRSAM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24666-1-AP-20UL
This item is not returnable.
View return policy
Description
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.Specifications
LRSAM1 | |
Polyclonal | |
Unconjugated | |
LRSAM1 | |
hTAL, LRSAM1, RIFLE, TAL, Tsg101 associated ligase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227738, 90678 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UWE0, Q80ZI6 | |
LRSAM1 | |
LRSAM1 Fusion Protein Ag20415 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |