Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Dechlor Tablets
Lovibond Replacement Reagents for CHECKIT Comparator: Dechlor Tablets, Range: 0.02 to 1mg/L, White, Composition: Sodium Nitrite 2.5 to 5%, non-hazardous components 90 to 100%, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Zincon/EDTA, pH Range: 6.1
Brand: Lovibond™ 512350BT
Code : NEW
Additional Details : CAS Number : 7632-00-0 Weight : 0.02760kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
Lovibond Replacement Reagents for CHECKIT Comparator: Dechlor Tablets, Range: 0.02 to 1mg/L, White, Composition: Sodium Nitrite 2.5 to 5%, non-hazardous components 90 to 100%, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Zincon/EDTA, pH Range: 6.1
Specifications
White | |
6.1 | |
Water analysis | |
0.02 to 1mg/L | |
Dechlor Tablet |
Sodium Nitrite 2.5 to 5%, non-hazardous components 90 to 100% | |
Tablet | |
Colorimeter and spectrophotometer | |
Zincon/EDTA |
Safety and Handling
- dechlor
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : sodium nitrate