Learn More
BioWhittaker™ Water for Cell Culture, Lonza™
Water for Injection (WFI) system for culture applications. BioWhittaker™ Water for Cell Culture is manufactured aseptically under cGMP conditions using a validated sterile filtration method.
POA
Description
Description
  • Method for the production of WFI includes:
  • Ultra filtration
  • Reverse osmosis
  • Mixed bed deionizers
  • Hot recirculating loop and cooled at point of use
  • System is validated and monitored for bioburden, endotoxin, total organic carbons and conductivity