missing translation for 'onlineSavingsMsg'
Learn More

Manutan Locker, perforated door, grey/dark grey, two tier, 1800mm x 300mm x 300mm

Brand:  Manutan A330154

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12895418

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Quality steel lockers with perforated doors and BioCote antibacterial technology.
  • Two tier
  • Dark grey
  • Dimensions (H x W x D): 1800mm x 300mm x 300mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers