missing translation for 'onlineSavingsMsg'
Learn More

Litmus, Indicator (pH 5-8), for titration, Honeywell Fluka™

Product Code. 15606040
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator (pH 5-8), for titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00000826

Specifications

CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula CHCl3
Grade Analytical Reagent
Packaging Glass Bottle
Quantity 500 g
Chemical Name or Material Litmus, Indicator (pH 5-8), for titration
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.