Learn More
Lithium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals
£25.02 - £599.99
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
PubChem CID | 433294 |
ChEBI | CHEBI:48607 |
SMILES | [Li+].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10187510
|
Thermo Scientific Acros
413270050 |
5 g | Glass Bottle |
£25.02
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328823
|
Thermo Scientific Acros
413271000 |
100 g | Plastic bottle |
£50.35
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10507942
|
Thermo Scientific Acros
413275000 |
500 g | Plastic bottle |
£160.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743744
|
Thermo Scientific Acros
413270025 |
2.5 kg | Plastic bottle |
£599.99
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
433294 | |
[Li+].[Cl-] |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
CHEBI:48607 |
Specifications
1382.0°C | |
7447-41-8 | |
Crystalline Powder and/or Crystals | |
ClLi | |
MFCD00011078 | |
15, 5584 | |
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone | |
[Li+].[Cl-] | |
42.39 | |
CHEBI:48607 | |
≥99% | |
1.0% max. (1 g, 105°C, 2 hrs) | |
2.0600g/mL |
605.0°C | |
White | |
99% min. (Argentometry) | |
LiCl | |
01,609; 02,246; 04,298; 05,677; 12,277; 16,194 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium(1+) chloride | |
433294 | |
42.38 | |
ACS Reagent | |
2.06 | |
Lithium chloride, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-212-3
missing translation for 'rtecsNumber' : OJ5950000
missing translation for 'tsca' : TSCA
- Lithium chloride, anhydrous
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Lithium chloride
RUO – Research Use Only