missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals

Product Code. 10665832
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
5 kg
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-85-3
Molecular Formula AlH4Li
MDL Number MFCD00011075
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445

Specifications

Melting Point 125.0°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 95%
Linear Formula LiAlH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Solubility Information Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES [Li+].[AlH4-]
IUPAC Name lithium(1+) alumanuide
Molecular Weight (g/mol) 37.95
Formula Weight 37.95
Percent Purity 95%
Density .9g/mL
Chemical Name or Material Lithium Aluminum hydride
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P402-Store in a dry place.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Lithium Aluminium Hydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'