missing translation for 'onlineSavingsMsg'
Learn More

Manutan Light duty anti-fatigue ribbed pattern mat, black 1520mm x 910mm

Brand:  Manutan A353544

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12867589

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

9. 4mm thickness anti-fatigue ribbed pattern for enhanced traction. Microcellular vinyl consists of millions of air bubbles providing basic worker comfort. All four sides are bevelled on mats (two sides on linear) and sealed to minimise tripping hazards (slip resistance R10 according to DIN 51130 and BG-rule BGR181). Free of toxic DOP with fire rating Class 2 according to NFPA Safety Code 101.
  • Black
  • Weight: 2.3kg/m2
  • Mat thickness: 9.4mm
  • Dimensions (L x W): 1520mm x 910mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers