Learn More
Invitrogen™ Levonorgestrel (LNG) Competitive ELISA Kit
Competitive ELISA
£606.00 - £3590.00
Specifications
Assay Sensitivity | 2.2 pg/mL |
---|---|
Description | Levonorgestrel (LNG) Competitive ELISA Kit |
Conjugate | HRP |
Detection Method | Fluorescence |
Product Type | ELISA Kit |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15924224
|
Invitrogen™
EIALNGC |
96 assays |
£606.00
Pack of 96 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17825217
|
Invitrogen™
EIALNGCX10 |
10 x 96 Assays |
£3590.00
Pack of 10 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
The Levonorgestrel (LNG) ELISA quantitates LNG in serum,plasma,urine,dried fecal extracts,saliva,milk,or cell culture medium.
Principle of the method
The Levonorgestrel (LNG) Competitive ELISA research-use-only kit is designed to quantitatively measure levonorgestrel present in serum,plasma,saliva,urine,dried fecal samples,milk,and tissue culture media samples. This kit measures total LNG in extracted serum,plasma,milk and fecal samples. A levonorgestrel stock solution is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. A levonorgestrel-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a rabbit polyclonal antibody to levonorgestrel to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound levonorgestrel-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Specifications
2.2 pg/mL | |
HRP | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
9% | |
RUO | |
-20°C | |
1 hr. 20 min. |
Levonorgestrel (LNG) Competitive ELISA Kit | |
Fluorescence | |
Fecal Extract, Milk, Plasma, Saliva, Serum, Supernatant, Urine, Water | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
9.1% | |
HRP | |
Reproductive Biology | |
Chemical | |
1 hr. 30 min. |