missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leuprolide
For cellular and molecular biology applications
£31.09 - £148.00
Specifications
Purification Method | HPLC |
---|---|
Common Name | Leuprorelin |
Format | Powder |
For Use With (Application) | Highly active luteinizing hormone releasing hormone (LHRH) agonist |
Molecular Weight (g/mol) | 1209.41 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15464569
|
Thermo Scientific Chemicals
J62147.M¬ |
1 mg | |||||||
15454569
|
Thermo Scientific Chemicals
J62147.#0 |
5 mg |
£148.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly active luteinizing hormone releasing hormone (LHRH) agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Powder | |
1209.41 | |
RUO - research use only |
Leuprorelin | |
Highly active luteinizing hormone releasing hormone (LHRH) agonist | |
Leuprolide | |
[Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH |
RUO – Research Use Only