Learn More
Greiner Bio-One Leucosep Polypropylene Round Bottom Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
Brand: Greiner Bio-One 163288
This item is not returnable.
View return policy
Description
- Enrichment directly from whole blood
- Simplified filling through porous barrier
- Rapid separation in 15 minutes at room temperature
- Removal of erythrocytes and granulocytes
- No recontamination with erythrocytes
- Service Plan
No
Specifications
12 mL | |
Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow | |
Aseptically Manufactured | |
Centrifuge Tube (Pre-filled) |
Polypropylene | |
Porous barrier | |
Round |