missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals

For Use in Battery Electrodes, Manufacturing of Pigments and Dyes, As Analytical Lab Reagent

£71.96 - £454.26

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331829
View Documents
Thermo Scientific Alfa Aesar
012483.22
100 g
£71.96
100g
Please to purchase this item. Need a web account? Register with us today!
11311829
View Documents
Thermo Scientific Alfa Aesar
012483.36
500 g
£324.62
500g
Please to purchase this item. Need a web account? Register with us today!
11321829
View Documents
Thermo Scientific Alfa Aesar
012483.A3
2 kg
£454.26
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Acid-insoluble matter 0.2%, Carbon compounds (as C) 0.04%, Cl 0.002%, NO3 0.02%, SO4 0.05%, Mn 5ppm, Ca 0.02%, Cu 0.05%, K 0.05%, Na 0.1%
Specifications

Chemical Identifiers

1309-60-0
239.20
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O2Pb
MFCD00011165
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
O=[Pb]=O

Specifications

290°C (decomposition)
Brown to Black
97.0% min.
MFCD00011165
Insoluble in water and alcohol. Soluble in acetic acid.
O=[Pb]=O
239.20
239.19
Odorless
Lead(IV) oxide
1309-60-0
Powder
O2Pb
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
YADSGOSSYOOKMP-UHFFFAOYSA-N
plumbanedione
14793
ACS
9.38
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c

H272-H302+H332-H350-H360Df-H373

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III

missing translation for 'einecsNumber' : 215-174-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(IV) oxide

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only