Learn More
Invitrogen™ LDLR Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHLDLRX5
Additional Details : Weight : 0.33820kg
Description
The Human LDLR ELISA quantitates Hu LDLR in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu LDLR. Principle of the method The Human LDLR solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation--the process of receptor-mediated endocytosis (1). Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits (2). It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells (3).Specifications
P01130 | |
8 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
3949 | |
<12% | |
Wash Buffer, SAV-HRP, Standard, Pre-coated 96 well plates, Adhesive Plate Covers, Stop Solution, Chromogen, Assay Diluent Concentrate, Biotinylated Detection Antibody | |
5 x 96 Tests | |
Virology | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
8.19 pg/mL to 2000 pg/mL | |
8 pg/mL to 2000 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA pair antibody detects human LDLR. Other species not determined yet. | |
Human | |
Colorimetric Microplate Reader | |
FH,FHC,LDLCQ2 | |
<10% | |
HRP | |
RUO | |
Plasma, 2.5 μ;L; Serum, 2.5 μ;L; Supernatant, 100 μ;L | |
FH, FHC, LDLCQ2 | |
1 hr. 20 min. |