missing translation for 'onlineSavingsMsg'
Learn More

Lauric acid, sodium salt, 98%, Thermo Scientific Chemicals

£35.38 - £362.81

Chemical Identifiers

CAS 629-25-4
Molecular Formula C12H23NaO2
Molecular Weight (g/mol) 222.30
MDL Number MFCD00041754
InChI Key BTURAGWYSMTVOW-UHFFFAOYSA-M
Synonym sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
PubChem CID 2735067
IUPAC Name sodium;dodecanoate
SMILES [Na+].CCCCCCCCCCCC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10552214
View Documents Promotion Details
Thermo Scientific Acros
413040050
5 g Glass bottle
£35.38
5g
Please to purchase this item. Need a web account? Register with us today!
10250031
View Documents
Thermo Scientific Acros
413040250
25 g Glass bottle
£125.63
25g
Please to purchase this item. Need a web account? Register with us today!
10615871
View Documents
Thermo Scientific Acros
413041000
100 g Glass bottle
£362.81
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-25-4
222.30
BTURAGWYSMTVOW-UHFFFAOYSA-M
2735067
[Na+].CCCCCCCCCCCC([O-])=O
C12H23NaO2
MFCD00041754
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
sodium;dodecanoate

Specifications

629-25-4
Authentic
C12H23NaO2
MFCD00041754
BTURAGWYSMTVOW-UHFFFAOYSA-M
sodium;dodecanoate
2735067
98%
Lauric acid, sodium salt, 99%
White to Yellow
98%
CH3(CH2)10COONa
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
[Na+].CCCCCCCCCCCC([O-])=O
222.30
222.3
Powder or Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 211-082-4

missing translation for 'rtecsNumber' : OF0700000

missing translation for 'tsca' : TSCA

Product Identifier
  • Lauric acid, sodium salt

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only