Learn More
OHAUS™ Large Vessel Carrier
OHAUS™ Large Vessel Carrier increases the amount of usable space for your samples.
Brand:  OHAUS™ 30400071
 View more versions of this product
Product Code. 15732489
  • £1355.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Tube racks and dimpled mats securely hold tubes in place
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Carrier Platform
24.00 in.
61 x 61 cm
61.00 cm
61.00 cm
Large
Stainless Steel
61.00 cm
24.00 in.
24.00 in.
Large Vessel Carrier, 61 X 61 cm