missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb Excitation Filter, 15 mm
LanthaScreen™ Tb Excitation Filter, 15 mm
Brand: Invitrogen™ PV00215
Code : 50
Additional Details : Weight : 0.01000kg
Description
Excitation filter (diameter = 15 mm) for use with the LanthaScreen™ Tb assays and products: 340 nm filter with a 30 nm bandpass.For use primarily with the Perkin Elmer EnVision™ plate reader. Perkin Elmer EnVision™ Empty Filter Holders are required to mount the filters (Perkin Elmer cat# 2100-8110, 10 empty barcoded filter holders for custom filters plus locking rings).
For assistance in setting up your instrument, please contact our Technical Support team in North America at +1 760 603 7200, x40266.
Specifications
Tb Excitation Filter | |
Microplate Reader | |
Pharma and biopharma, target and lead identification and validation, protein biology, drug discovery and development, kinase biology, biochemical kinase assays, proteasome biology, biochemical proteasome assays, lanthascreen™ kinase activity assays |
For Research Use Only. Not for use in diagnostic procedures.