Learn More
Labtek™ Liquid Detergent
Labtek liquid is a free-rinsing liquid alkaline detergent, for cleaning laboratory instruments, glassware, ceramics and plastics
Brand: Teknon TK120/5
Code : 04
Additional Details : CAS Number : 68187-76-8 Weight : 5.50000kg
Description
Labtek liquid is a free-rinsing liquid alkaline detergent, for cleaning laboratory instruments, glassware, ceramics and plastics. Labtek liquid is free from enzymes and sodium hypochlorite. Used in a 2% to 10% solution it rapidly removes oil, grease, tar, silicones, inorganic soils, polyacrylate and PVA resins, blood, proteins and a wide range of other organic soils from ferrous metals, glassware, plastics and ceramics. Can be used manually or in an ultrasonic bath where heating the solution improves cleaning efficiency. Not suitable for use on non-ferrous metals and polycarbonate.- Multi purpose
- Powerful wetting agent
- Ultrasonic compatible
- Free rinsing The product is highly suitable for use in pharmaceutical, scientific and industrial laboratories where detergent residues are an issue.
Specifications
Labtek Liquid Detergent | |
Suitable for use in pharmaceutical, scientific and industrial laboratories where detergent residues are an issue | |
1 Ea. | |
5 L |
Safety and Handling
- Labtek Liquid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P302+P334-IF ON SKIN: Immerse in cool water/wrap in wet bandages.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.