Learn More
3M™ 4432+ Lab Coats Product Code.: 15388756
3M™ 4432+ Lab Coats
Product Code. 15388756
X-Large
Click to view available options
Size:
X-Large
2X-Large
Product Code. 15388756
Brand: 3M™ 7100044082
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to help protect the wearer against light splashing with non-hazardous liquids and dust. 3M™ 4432+ Lab Coats are made from super-breathable SSMMS non-woven for effective release of body heat and moisture.
  • Anti-static
  • Front zip closing
  • Low lint properties
  • Reduced heat stress
  • Knitted cuffs and collared
  • Internal pocket on the wearer's left-hand side
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Polypropylene, Polyethylene Laminate
Color Cool White
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Seam Type Serged
Certifications/Compliance CE Category 1
Packaging Type Single Pack
Product Title
3M™ 4432+ Lab Coats >