Learn More
Labconco™ Protector™ Acid and Standard Storage Cabinets Shelf Kits
Shelf Kits provide additional storage space in Labconco Protector Standard and Acid Base Cabinets.
£399.00
Product Code | Brand | For Use With (Equipment) | Includes | Width (English) Exterior | Width (Metric) Exterior | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Width (English) Exterior | Width (Metric) Exterior | Price | Quantity & Availability | ||
16348705
|
Labconco™
9919000 |
Protector™ Acid storage cabinet | Epoxy Coated Steel shelf, Acid-resistant Polyethylene tray and mounting hardware | 48 in. | 121.92 cm |
£399.00
Each |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Interior Shelf Kit provides additional storage space inside Labconco Protector Standard Storage Cabinets. Includes powder-coated steel shelf and mounting hardware. Shelf may be mounted at one of three heights: 13.0 in., 15.25 in., 17.0 in. (9.5 in., 11.25 in. and 13.0 in. for ADA height cabinets).Specifications
Shelf Kit |