missing translation for 'onlineSavingsMsg'
Learn More

Estrogen Receptor Ab-17 Rabbit Polyclonal Antibody, Epredia™

Epredia™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia RB-1521-P0

Code : LD

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12603017

  • £149.00 / 100µL
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ER gene consists of more than 140kb of genomic DNA divided into 8 exons, being translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Estrogen Receptor Ab-17
Polyclonal
Unconjugated
A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein
100 μL
Steroids Hormones and Growth Factors
Bovine, Human, Mouse, Porcine, Rabbit, Rat
IgG
Immunoprecipitation, Western Blot
1mg/mL
Rabbit
Purified with BSA and azide
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers