missing translation for 'onlineSavingsMsg'
Learn More

Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.

Brand:  Epredia MS-235-P1

Code : LD

Additional Details : Weight : 0.00190kg

 View more versions of this product

Product Code. 12660657

  • £393.00 / 500µL
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Mouse

Clone: 2B1.5

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage in lung or fetus

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Collagen II Ab-2
Monoclonal
200μg/mL
Mouse
500 μL
Extracellular Matrix
Bovine, Chicken, Human, Mouse, Rat
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2B1.5
Unconjugated
A purified preparation of lathyritic type II collagen from embryonic chicken sternum
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers