Learn More
Berkshire™ Lab-Tips™ Low TOC Swab Large
Berkshire Lab-Tips™ Low TOC Swab Large knitted polyester swab features a unique semi-flexible internal paddle for greater surface coverage.
Brand:  Berkshire™ LTP125TOC.25
Product Code. 16468935
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Specifically designed for Total Organic Carbon analysis and TOC tests in low-TOC environment
  • Critically low particles, fibers, ions and extractables
  • Excellent chemical resistance
  • High abrasion resistance while soft and non-abrasive
  • No contaminating adhesives and good sorbency
Specifications
Specifications
20 swabs/pack; 25 packs/case
TOC Swab
Polyester
3.94 in.
100mm
Large