missing translation for 'onlineSavingsMsg'
Learn More

L-Malic acid, ≥99%, Thermo Scientific Chemicals

£264.00 - £483.00

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2 s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2 S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15815178
View Documents
Thermo Scientific
J18700.36
500 g Plastic bottle
£264.00
500g
Please to purchase this item. Need a web account? Register with us today!
15835318
View Documents
Thermo Scientific
J18700.A1
1 kg Plastic bottle
£483.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

97-67-6
134.087
BJEPYKJPYRNKOW-REOHCLBHSA-N
222656
(2 S)-2-hydroxybutanedioic acid
C4H6O5
MFCD00064213
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2 s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
CHEBI:30797
C(C(C(=O)O)O)C(=O)O

Specifications

97-67-6
C4H6O5
1723541
BJEPYKJPYRNKOW-REOHCLBHSA-N
(2 S)-2-hydroxybutanedioic acid
222656
134.09
1.6
Crystalline Powder
L-Malic acid
100°C to 106°C
MFCD00064213
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2 s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:30797
≥99%
>110°C
1.6
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'einecsNumber' : 202-601-5

missing translation for 'rtecsNumber' : ON7175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only