missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99%

£44.50 - £419.00

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10625042
View Documents Promotion Details
Thermo Scientific Chemicals
173730250
25 g Plastic Bottle
£44.50
25g
Please to purchase this item. Need a web account? Register with us today!
10124050
View Documents Promotion Details
Thermo Scientific Chemicals
173731000
100 g Plastic Bottle
£127.00
100g
Please to purchase this item. Need a web account? Register with us today!
10044093
View Documents
Thermo Scientific Chemicals
173735000
500 g Plastic Bottle
£419.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39665-12-8
164.21
HZRUTVAFDWTKGD-XZNNNFJINA-N
16211825
C6H16N2O3
MFCD00151035
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
O.NCCCC[C@H](N)C(O)=O

Specifications

212.0°C to 214.0°C
39665-12-8
White to Yellow
10ppm max.
10ppm max.
0.5% max. (TLC)
H2N(CH2)4CH(NH2)CO2H·H2O
04, 435
200ppm max.
95% min. (c=5, H2O, 430nm) 1cm cell
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
10 to 11.5%
O.NCCCC[C@H](N)C(O)=O
+ 26.25 (20.00°C c=8,6N HCl)
16211825
99%
L(+)-Lysine monohydrate
200ppm max.
9 to 10.5 (2.5% soln. at 25°C)
300ppm max.
Authentic
99%
C6H16N2O3
MFCD00151035
+ 26.25
14, 5636
0.1% max.
Solubility in water: 1500g/L (20°C). Other solubilities: insoluble in common neutral solvents
HZRUTVAFDWTKGD-XZNNNFJINA-N
(2S)-2,6-diaminohexanoic acid hydrate
164.21
164.21
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • L(+)-Lysine monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only