missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 57282-49-2 |
---|---|
Molecular Formula | C8H18N2O4 |
Molecular Weight (g/mol) | 206.24 |
MDL Number | MFCD00039069 |
InChI Key | RRNJROHIFSLGRA-JEDNCBNOSA-N |
Synonym | l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz |
PubChem CID | 104152 |
IUPAC Name | acetic acid;(2S)-2,6-diaminohexanoic acid |
SMILES | CC(O)=O.NCCCC[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15294874
|
Thermo Scientific Acros
451630250 |
25 g |
£23.62
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15214884
|
Thermo Scientific Acros
451631000 |
100 g |
£64.22
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15224884
|
Thermo Scientific Acros
451635000 |
500 g |
£226.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57282-49-2 | |
206.24 | |
RRNJROHIFSLGRA-JEDNCBNOSA-N | |
104152 | |
CC(O)=O.NCCCC[C@H](N)C(O)=O |
C8H18N2O4 | |
MFCD00039069 | |
l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz | |
acetic acid;(2S)-2,6-diaminohexanoic acid |
Specifications
57282-49-2 | |
C8H18N2O4 | |
l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz | |
CC(O)=O.NCCCC[C@H](N)C(O)=O | |
206.24 | |
206.24 | |
L-Lysine acetate |
97% | |
MFCD00039069 | |
RRNJROHIFSLGRA-JEDNCBNOSA-N | |
acetic acid;(2S)-2,6-diaminohexanoic acid | |
104152 | |
97% |
Safety and Handling
Product Identifier
- L-Lysine acetate
RUO – Research Use Only