Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint
Ideal for use with rotary evaporators
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Equipment) | Rotary evaporator |
Neck Style | Single |
Description
Ideal for use with rotary evaporators
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- Single-neck flask with standard taper outer joint
- Designed for easy recovery of reaction products
- Total contents are accessible with a spatula or policeman
- Lower portion fits standard heating mantles
- Even wall thickness and minimum runout
Specifications
Borosilicate Glass | |
Rotary evaporator | |
Single |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint