missing translation for 'onlineSavingsMsg'
Learn More

KLHL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen KLHL2
Concentration 0.22 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16853094
View Documents
Proteintech
14449-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16843094
View Documents
Proteintech
14449-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.
Specifications

Specifications

KLHL2
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Mouse, Rat, Human
F1LZF0, O95198, Q8JZP3
11275, 290692, 77113
KLHL2 Fusion Protein Ag5950
Primary
-20°C
KLHL2
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ABP KELCH, Actin binding protein Mayven, Kelch like protein 2, KLHL2, MAV, MAYVEN
Klhl2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications