Learn More
Ecolab™ Klerwipe™ 70/30 IPA
Sterile low particulate wipes impregnated with 70% isopropyl alcohol blended with deionised water
Brand: Ecolab™ 3079390
Code : NEW
Additional Details : CAS Number : 67-63-0 Weight : 6.50000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- The alcohol blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
- A sterile ready-to-use wipe for small surfaces available in either a resealable pouch featuring Klertear on outer wrap, or in a tub
Specifications
Pouch Wipe in VHP | |
200 x 200 mm | |
20 x 15 Wipes | |
70% Isopropyl Alcohol |
Safety and Handling
- Klerwipe 70/30 IPA
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl Alcohol