missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ 70/30 Denatured Ethanol

Sterile low particulate wipes impregnated with 70% denatured ethanol blended with deionised water

Brand:  Ecolab™ 3079190

Code : NEW

Additional Details : CAS Number : 64-17-5 Weight : 6.50000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 15376108

  • £855.53 / Pack of 15
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • The alcohol blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
  • A sterile ready-to-use wipe for small surfaces available in either a resealable pouch, featuring Klertear on outer wraps, or in a tub
Specifications

Specifications

Low Particulate Tub Wipe
200 x 230 mm
15 x 100 Wipes
70% Denatured Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Klerwipe 70/30 Denatured Ethanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers