Learn More
Ecolab™ Klerwipe™ 70/30 Denatured Ethanol
Sterile low particulate wipes impregnated with 70% denatured ethanol blended with deionised water
Brand: Ecolab™ 3079160
283.16 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 3.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- The alcohol blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
- A sterile ready-to-use wipe for small surfaces available in either a resealable pouch, featuring Klertear on outer wraps, or in a tub
Specifications
Low Particulate Pouch Wipe | |
200 x 200 mm | |
20 x 15 Wipes | |
70% Denatured Ethanol |
Safety and Handling
- Klerwipe 70/30 Denatured Ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol