Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
£159.12
Specifications
Certifications/Compliance | ASTM, E1406, Type I |
---|---|
Material | Borosilicate Glass |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15332324
|
DWK Life Sciences
270504000 |
4 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Heavy walled
- Necks tooled for uniform stopper fit with a thick flat flange
- Without side tabulation
- Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
ASTM, E1406, Type I | |
Borosilicate Glass |