Learn More
DWK Life Sciences Kimble™ Aluminum Seal Caps
With PTFE-faced rubber septa
Specifications
For Use With (Application) | Multiple injection |
---|---|
Material | Aluminum |
Neck Finish | 11 |
Description
- Moderate to good resealability
- One piece
- PTFE-faced red rubber septa
- For general GC applications
- Moderate resealability
- Not recommended for multiple injections
- PTFE-faced natural rubber septa
- Good resealability
- Multiple injections
Specifications
Multiple injection | |
Aluminum | |
11 |