missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Gooch Filtering Crucibles

Dry precipitates to constant weight at 110°C (230°F)

Brand:  DWK Life Sciences 28260-301

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 15399933

  • £530.00 / Pack of 12
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

  • At higher temperatures, heat in electric furnace
  • Do not remove crucibles until temperature is reduced to about 250°C (482°F)
  • Fritted disc
  • Avoid exposure to sudden temperature changes
  • Coarse (C), medium (M), and fine (F) porosities

Specifications

Specifications

USP Type I, ASTM E438, Type I, Class A
3cm
For collecting, drying and weighing precipitates and samples to be analyzed using gravimetric analysis
30 mL
Gooch Filtering Crucible
With Fritted discs
Borosilicate Glass
250°C
5cm
Coarse
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers