missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDC3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24989-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the KHDC1 family, members of which contain an atypical KH domain that may not bind RNA like canonical KH domains. This gene is specifically expressed in the oocytes, and recent studies suggest that it may function as a regulator of genomic imprinting in the oocyte. Mutations in this gene are associated with recurrent biparental complete hydatidiform mole.Specifications
KHDC3L | |
Polyclonal | |
Unconjugated | |
KHDC3L | |
C6orf221, ECAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
154288 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q587J8 | |
KHDC3L | |
KHDC3L Fusion Protein Ag21790 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |