missing translation for 'onlineSavingsMsg'
Learn More

Kerosene Blank, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

£54.85 - £124.00

Specifications

Chemical Name or Material Kerosene Blank
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Type Certified Reference Material
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11450269
View Documents
SPEX Certiprep
KER-BLK
500 mL Triple-leached LDPE bottle
£54.85
500mL
Please to purchase this item. Need a web account? Register with us today!
11460269
View Documents
SPEX Certiprep
KER-BLK-G
1 gal. Triple-leached LDPE bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Kerosene Blank
  • Organometallic Oil Standard

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Kerosene Blank
With Certificate of Analysis.
Certified Reference Material
Kerosene
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Petrochemical Testing
Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Kerosine (petroleum)

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.