Learn More
SI Analytics™ Karl Fischer Titration Vessel
SI Analytics™ Karl Fischer Titration Vessel is constructed of DURAN™ glass and comes equipted with screw threads, a titration tip, and more.
Specifications
Volume (Metric) 30 to 150 mL
View More Specs
Description
Description
  • Includes a removable head made of polypropylene/PTFE
  • Includes 1 drilling NS 19, NS 14.5, NS 7.5 and 3 drillings with screw threads
  • Includes a titration tip, moisture trap and weighing funnel
Specifications
Specifications
30 to 150 mL