Learn More
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 5150 H24
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
£1695.00
Specifications
Pump Type Force
View More Specs
Products 1
Product Code Brand Frequency Price Quantity & Availability  
Product Code Brand Frequency Price Quantity & Availability  
11376671
View Documents Promotion Details
Fisherbrand™
211213100
50 Hz
£1695.00
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
Description
Description
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters
  • High temperature and low level cut-outs
  • Compact footprint maximizes bench space
  • Includes external circulation plumbing with 8-mm and 12-mm fittings and supply and return clamps
  • Work area cover included on all heated baths
  • High quality stainless steel baths
  • All bath drains are located on the front of the unit with an integrated valve
Specifications
Specifications
Force