Learn More
Isopropylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£19.90 - £183.00
Chemical Identifiers
CAS | 76347-13-2 |
---|---|
Molecular Formula | C9H19BO2 |
Molecular Weight (g/mol) | 170.059 |
MDL Number | MFCD05663856 |
InChI Key | MECCSFDHAVAAAW-UHFFFAOYSA-N |
Synonym | 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 12727711 |
IUPAC Name | 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496737
|
Thermo Scientific Alfa Aesar
H33861.03 |
1 g |
£19.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406747
|
Thermo Scientific Alfa Aesar
H33861.06 |
5 g |
£48.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416747
|
Thermo Scientific Alfa Aesar
H33861.14 |
25 g |
£183.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsopropylboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
76347-13-2 | |
170.059 | |
MECCSFDHAVAAAW-UHFFFAOYSA-N | |
12727711 | |
B1(OC(C(O1)(C)C)(C)C)C(C)C |
C9H19BO2 | |
MFCD05663856 | |
2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane |
Specifications
76347-13-2 | |
C9H19BO2 | |
MFCD05663856 | |
Slightly soluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C(C)C | |
170.059 | |
170.06 | |
Isopropylboronic acid pinacol ester |
115°C (60 mmHg) | |
1.409 | |
2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
MECCSFDHAVAAAW-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane | |
12727711 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.