Learn More
Isopropyl myristate, 96%, Thermo Scientific Chemicals
£14.57 - £97.59
Chemical Identifiers
CAS | 110-27-0 |
---|---|
Molecular Formula | C17H34O2 |
Molecular Weight (g/mol) | 270.45 |
MDL Number | MFCD00008982 |
InChI Key | AXISYYRBXTVTFY-UHFFFAOYSA-N |
Synonym | isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester |
PubChem CID | 8042 |
IUPAC Name | propan-2-yl tetradecanoate |
SMILES | CCCCCCCCCCCCCC(=O)OC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10710682
|
Thermo Scientific Acros
412850250 |
25 g | Glass bottle |
£14.57
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687191
|
Thermo Scientific Acros
412851000 |
100 g | Glass bottle |
£20.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676504
|
Thermo Scientific Acros
412850010 |
1 kg | Glass bottle |
£97.59
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-27-0 | |
270.45 | |
AXISYYRBXTVTFY-UHFFFAOYSA-N | |
8042 | |
CCCCCCCCCCCCCC(=O)OC(C)C |
C17H34O2 | |
MFCD00008982 | |
isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
propan-2-yl tetradecanoate |
Specifications
0.5mg KOH/g max. | |
0.8500g/mL | |
>110°C | |
95% min. (GC) | |
CH3(CH2)12COOCH(CH3)2 | |
MFCD00008982 | |
0.85 | |
AXISYYRBXTVTFY-UHFFFAOYSA-N | |
propan-2-yl tetradecanoate | |
8042 | |
96% | |
Isopropyl myristate, 96% |
110-27-0 | |
167.0°C (9.0 mmHg) | |
Authentic | |
C17H34O2 | |
1.4340 to 1.4360 | |
15, 5261 | |
isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
CCCCCCCCCCCCCC(=O)OC(C)C | |
270.45 | |
270.45 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 203-751-4
missing translation for 'rtecsNumber' : XB8600000
missing translation for 'tsca' : TSCA
- Isopropyl myristate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.