missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyltriethoxysilane, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in a wide range of applications including silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent, silane cross linking agent, silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent, silane reinforcer, silylating agent, silane reducing agent, silyl building blocks and synthons.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
It is used in a wide range of applications including silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent, silane cross linking agent, silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent, silane reinforcer, silylating agent, silane reducing agent, silyl building blocks and synthons.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Boiling Point | 165°C (400mmHg) |
Melting Point | 190°C to 191°C |
Physical Form | Liquid |
Quantity | 25 g |
Linear Formula | (CH3)2CHCH2Si(OC2H5)3 |
Beilstein | 1750049 |
Solubility Information | Insoluble in water. |
Formula Weight | 220.38 |
Percent Purity | 95% |
Refractive Index | 1.4505 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Isobutyltriethoxysilane, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction