missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, purified, Honeywell Fluka™

Product Code. 15631130
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purified,≥95%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Quantity 6 x 1 kg
UN Number NONH for all modes of transport
Formula Weight 159.69
Percent Purity ≥95%
Grade purified
Chemical Name or Material Iron(Iii) Oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'