missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Chloride, Honeywell
Reagent Grade, 97%
£29.27 - £502.02
Chemical Identifiers
CAS | 7705-08-0 |
---|---|
Molecular Formula | Cl3Fe |
Molecular Weight (g/mol) | 162.20 |
MDL Number | MFCD00011005 |
InChI Key | RBTARNINKXHZNM-UHFFFAOYSA-K |
IUPAC Name | iron(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15631510
|
Honeywell Research Chemicals
157740-5G |
5 g | N/A | ||||||
15691500
|
Honeywell Research Chemicals
157740-100G |
100 g | N/A | ||||||
15601510
|
Honeywell Research Chemicals
157740-1KG |
1 kg | N/A | ||||||
15611510
|
Honeywell Research Chemicals
157740-2.5KG |
2.5 kg | N/A | ||||||
15621510
|
Honeywell Research Chemicals
157740-20KG |
20 kg | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7705-08-0 | |
162.20 | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3Fe | |
MFCD00011005 | |
iron(3+) trichloride |
Specifications
7705-08-0 | |
FeCl3 | |
1773 | |
[Cl-].[Cl-].[Cl-].[Fe+3] | |
162.20 | |
97% | |
1mmHg (194°C) | |
Iron(Iii) Chloride |
Cl3Fe | |
MFCD00011005 | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
iron(3+) trichloride | |
162.2g/mol | |
Reagent | |
Vapor Density: 5.61 (vs air) |
Safety and Handling
P280-P301 + P312 + P330-P305 + P351 + P338 + P310
H290-H302-H315-H318
missing translation for 'einecsNumber' : 231-729-4
missing translation for 'rtecsNumber' : LJ9100000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Iron(III) chloride
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.