missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10311071
Click to view available options
Quantity:
5 g
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64.0°C
Color Blue or Blue-Green or Green
Physical Form Crystals
Quantity 3 kg
Assay Percent Range 99+%
Linear Formula FeSO4·7H2O
Fieser 01,393; 04,237; 05,308
Merck Index 15,4084
Solubility Information Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol
IUPAC Name iron(2+);sulfate;heptahydrate
Formula Weight 278.01
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.