missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ PSF Syringe Filters, 0.45 μm
Syringe filters specifically designed for ion chromatography applications.
Brand: Cytiva (formerly Pall Lab) AP-4585
111.40 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Optimized to provide the most consistent results when analyzing ionic species
Specifications
PES | |
Non-sterile | |
Polypropylene | |
50/Pk. | |
25 mm |
0.45 μm | |
3.9 cm2 | |
200/Cs. | |
100°C |