Learn More
Iodoform, 99%, Thermo Scientific Chemicals
Description
Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.
Solubility
Soluble in water, ether, acetic acid, ethanol, chloroform, ether, glycerol, carbon disulfide, olive oil, benzene and acetone. Slightly soluble in petroleum ether.
Notes
Light sensitive. Incompatible with strong bases, alkali metals, trong oxidizing agents and magnesium.

Specifications
Specifications
Melting Point | 118°C to 123°C |
Density | 4.008 |
Flash Point | None |
Odor | Unpleasant |
Quantity | 250 g |
Beilstein | 1697010 |
Sensitivity | Light sensitive |
Merck Index | 14,5033 |
Solubility Information | Soluble in water,ether,acetic acid,ethanol,chloroform,ether,glycerol,carbon disulfide,olive oil,benzene and acetone. Slightly soluble in petroleum ether. |
Formula Weight | 393.73 |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.