Learn More
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 626-62-0 |
---|---|
Molecular Formula | C6H11I |
Molecular Weight (g/mol) | 210.06 |
MDL Number | MFCD00003826 |
InChI Key | FUCOMWZKWIEKRK-UHFFFAOYSA-N |
Synonym | cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i |
PubChem CID | 12289 |
IUPAC Name | iodocyclohexane |
SMILES | IC1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11436626
|
Thermo Scientific Alfa Aesar
B24840.14 |
25 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446626
|
Thermo Scientific Alfa Aesar
B24840.22 |
100 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456626
|
Thermo Scientific Alfa Aesar
B24840.36 |
500 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
Solubility
Soluble in ethanol, ether, acetone.
Notes
Light sensitive. Store away from oxidizing agent, light.
Chemical Identifiers
626-62-0 | |
210.06 | |
FUCOMWZKWIEKRK-UHFFFAOYSA-N | |
12289 | |
IC1CCCCC1 |
C6H11I | |
MFCD00003826 | |
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i | |
iodocyclohexane |
Specifications
626-62-0 | |
180°C | |
C6H11I | |
MFCD00003826 | |
Light sensitive | |
Soluble in ethanol,ether,acetone. | |
IC1CCCCC1 | |
210.06 | |
210.06 | |
Iodocyclohexane, stabilized with copper |
1.625 | |
71°C (159°F) | |
1.5475 | |
1900797 | |
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i | |
FUCOMWZKWIEKRK-UHFFFAOYSA-N | |
iodocyclohexane | |
12289 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 210-957-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.