missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Split Liner
Specifically designed to ensure a highly inert GC pathway for basic compounds
Brand: Restek™ 20782-211.5
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Split liner design with base-deactivated wool packing material.
Base-deactivation process:
- Bonds basic functional groups to the analytical surface to reduce adsorption and priming effects
- I.D. 4mm
- Improves peak shape, response and reproducibility when compared to standard deactivation
- Base-deactivated wool prevents the vaporized sample from contacting the adsorptive hot metal inlet disc surface
Specifications
Base Deactivated Wool | |
5/Pk. | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |
Split Base Deactivated Straight Inlet Liner | |
7.85 cm | |
0.63 cm | |
Borosilicate Glass |