missing translation for 'onlineSavingsMsg'
Learn More

Powdered Vinyl Examination Gloves for First Aid Kit

Product Code. 12930465
This item is not returnable. View return policy
This item is not returnable. View return policy

Keep Powdered Vinyl Examination Gloves for First Aid Kit refills on hand per the Health and Safety ACOP L74 regulation for contingency supplies in case of emergency.

With reference to first aid kit refills, the Health and Safety ACOP L74 states “Sufficient supplies should be held as back-up on site”.

Specifications

Disposable Single-use
Grade Medical quality AQL 1.5
Latex Free Yes
Material Vinyl
Powdered Powdered
Product Type Gloves
Sterility Non-sterile
Style Powdered
For Use With (Application) Medical and electronics
For Use With (Equipment) First aid kit
Inspected for Medical Use Yes
Packaging Type 6 pairs per pack
Quantity 6Pair
Weight (Metric) 1.0 kg
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'