missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid solution, 2M EU Pharmacopoeia, Fisher Chemical™

Product Code. 12970854
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 7647-01-0, 7732-18-5
Concentration 2M
Packaging Bottle
Quantity 1000 mL
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313  
ChEBI CHEBI:23116
Chemical Name or Material Hydrochloric Acid solution
Show More Show Less
compliance-icons
Product Identifier
  • hydrochloric acid solution EU pharmacopeia grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'