missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, 0.05M, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Concentration | 0.05M |
Vapor Pressure | 190 mmHg (25°C) |
Quantity | 500 mL |
UN Number | UN1789 |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46g/mol |
Chemical Name or Material | Hydrochloric acid solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'